Please login to your online account to display your discounted pricing

Acetyl-β-methylcholine bromide, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Acetyl-β-methylcholine bromide
Name Note 99%
CAS 333-31-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (on dried substance)
Assay Percent Range 99%
Molecular Formula C8H18BrNO2
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Br
Formula Weight 240.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC226970050

  SDS
thermo fisher scientific
226970050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC226970250

  SDS
thermo fisher scientific
226970250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (on dried substance)
Physical Form Crystalline Powder
Formula Weight 240.14
Quantity 5g
Color White to off-white
Beilstein 04, III, 758
Merck Index 10, 5793
EINECS Number 206-372-2
TSCA TSCA
Melting Point 149° to 153°C
CAS Max % 100.0
Chemical Name or Material Acetyl-β-methylcholine bromide
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Br
MDL Number MFCD00011816
CAS 333-31-3
CAS Min % 98.5
Loss on Drying 1% max.
Molecular Formula C8H18BrNO2
Packaging Glass bottle
Synonym Methacholine bromide
Health Hazard 1 HYGROSCOPIC
Safety and Handling

HYGROSCOPIC