Please login to your online account to display your discounted pricing

Acetyl chloride, ACS reagent, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC447170010

 View more versions of this product

Catalog No. AC447170010

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Acetyl chloride
CAS 75-36-5
Health Hazard 1 Danger
Packaging Glass bottle
Purity Grade ACS Reagent Grade
Molecular Formula C2H3ClO
Linear Formula CH3COCl
Formula Weight 78.5
Density 1.1000g/mL
Quantity 1L

Specifications

Chemical Name or Material Acetyl chloride
CAS 75-36-5
Purity Grade ACS Reagent Grade
Molecular Formula C2H3ClO
Linear Formula CH3COCl
Formula Weight 78.5
Accessories

Danger

SDS & Safety Documents