Please login to your online account to display your discounted pricing

Acetyl chloride, ACS reagent, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC447172500

 View more versions of this product

Catalog No. AC447172500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 78.5
Quantity 250mL
Density 1.1000g/mL
Chemical Name or Material Acetyl chloride
Packaging Glass bottle
Molecular Formula C2H3ClO
CAS 75-36-5
Health Hazard 1 Danger
Linear Formula CH3COCl

Specifications

Chemical Name or Material Acetyl chloride
CAS 75-36-5
Purity Grade ACS Reagent Grade
Molecular Formula C2H3ClO
Linear Formula CH3COCl
Formula Weight 78.5
Accessories

Danger

SDS & Safety Documents