missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl chloride, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 43262AD

 View more versions of this product

Catalog No. AA43262AD



Specifications

Specifications

Acetyl chloride
50mL
≈99%
MFCD00000719
WETWJCDKMRHUPV-UHFFFAOYSA-N
acetyl chloride
6367
78.5
1.104
75-36-5
C2H3ClO
acetic acid chloride
CC(=O)Cl
78.495
CHEBI:37580
Liquid

Chemical Identifiers

75-36-5
78.495
WETWJCDKMRHUPV-UHFFFAOYSA-N
6367
acetyl chloride
C2H3ClO
MFCD00000719
acetic acid chloride
CHEBI:37580
CC(=O)Cl
Safety and Handling

Safety and Handling

EINECSNumber : 200-865-6

Documents