Please login to your online account to display your discounted pricing

Acetyl chloride, 99+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Acetyl chloride
CAS 75-36-5
Molecular Formula C2H3ClO
Linear Formula CH3COCl
Formula Weight 78.5
MDL Number MFCD00000719
UN Number UN1717
Beilstein 605303
Merck Index 14,85
Synonym Ethanoyl chloride; Acetic chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA43262K2

  SDS
alfa aesar
43262K2
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Acetyl chloride
CAS 75-36-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314;
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETYL CHLORIDE
RTECS Number AO6390000
TSCA TSCA
Refractive Index 1.3890
Physical Form Liquid
Assay ≥99%
Molecular Formula C2H3ClO
Linear Formula CH3COCl
Formula Weight 78.5
MDL Number MFCD00000719
UN Number UN1717
Beilstein 605303
Merck Index 14,85
Synonym Ethanoyl chloride; Acetic chloride
Density 1.104
Color Colorless
Boiling Point 50° to 52°C
Melting Point −112°C
Quantity 1L
Flash Point 4°C (39°F)
Odor Pungent
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314;
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.