missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl chloride, 98%, ACROS Organics™

Manufacturer:  Acros Organics 151270010

 View more versions of this product

Catalog No. AC151270010


 

Specifications

Specifications

Acetyl chloride
Authentic
1.3876 to 1.3896
98%
1.1
CH3COCl
01,11; 04,5; 05,4; 07,3; 11,4
1.1000g/mL
WETWJCDKMRHUPV-UHFFFAOYSA-N
CC(=O)Cl
51.0°C
78.495
CHEBI:37580
4°C
Liquid
75-36-5
Glass bottle
Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol
98%
C2H3ClO
02, 173
15, 80
Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol
Undesignated
acetyl chloride
-112.0°C
6367
1L
78.5

Chemical Identifiers

75-36-5
78.495
6367
C2H3ClO
WETWJCDKMRHUPV-UHFFFAOYSA-N
CHEBI:37580
Safety and Handling

Safety and Handling

Danger

SDS Documents