missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 387741000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetyl chloride | |
1M solution in dichloromethane | |
5.44,93.36 | |
C2H3ClO | |
MFCD00000719 | |
1.3000g/mL | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.5 | |
1.3 | |
Liquid |
Colorless | |
75-36-5,75-09-2 | |
6.64,94.57 | |
CH3COCl | |
0.9 to 1.1M | |
15, 80 | |
Solubility in water: reacts with water. | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
AcroSeal™ Glass bottle | |
100 mL |
RUO – Research Use Only