missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl bromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1248522

 View more versions of this product

Catalog No. AAA1248522



Description

Description

Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.

Solubility
Miscible with ether, chloroform, benzene and acetone.

Notes
Incompatible with strong oxidizing agents, strong bases and alcohols
Specifications

Specifications

Acetyl Bromide
°C to 96°C
75°C to 77°C
≥98%
1.45
MFCD00000114
UN1716
Moisture sensitive
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
FXXACINHVKSMDR-UHFFFAOYSA-N
acetyl bromide
10482
≥98%
506-96-7
1.660
>75°C (167°F)
C2H3BrO
CH3COBr
100 g
1697546
14,82
Miscible with ether,chloroform,benzene and acetone.
CC(=O)Br
122.949
122.95
Liquid

Chemical Identifiers

506-96-7
122.949
FXXACINHVKSMDR-UHFFFAOYSA-N
10482
CC(=O)Br
C2H3BrO
MFCD00000114
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
acetyl bromide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETYL BROMIDE

EINECSNumber : 208-061-7

RTECSNumber : AO5955000

TSCA : Yes

storageNote1 : Moisture Sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only