missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl bromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 159405000

 View more versions of this product

Catalog No. AC159405000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetyl bromide
506-96-7
100.0
Colorless to Yellow
75°C to 77°C
Authentic
Glass bottle
1.4485 to 1.4505
MFCD00000114
02, 174
1.66
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
FXXACINHVKSMDR-UHFFFAOYSA-N
acetyl bromide
10482
98%
98%
97.5
-96°C
1.6600g/mL
75°C
97.5% min. (Argentometry)
C2H3BrO
CH3COBr
500 mL
06,9; 13,208
15, 77
Solubility in water: reacts. Other solubilities: reacts with alcohol and acetone,soluble in ether,chloroform and benzene
CC(=O)Br
122.95
122.95
Fuming Liquid

Chemical Identifiers

506-96-7
122.95
FXXACINHVKSMDR-UHFFFAOYSA-N
10482
CC(=O)Br
C2H3BrO
MFCD00000114
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
acetyl bromide
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 208-061-7

RTECSNumber : AO5955000

TSCA : TSCA

Recommended Storage : May darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only