Please login to your online account to display your discounted pricing

Acetyl-beta-methylcholine chloride, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC102610250

 View more versions of this product

Catalog No. AC102610250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 195.69
Quantity 25g
Merck Index 15, 6012
CAS Max % 100.0
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Cl
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Chemical Name or Material Acetyl-β-methylcholine chloride
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H18ClNO2
Packaging Glass bottle
MDL Number MFCD00011817
CAS 62-51-1
CAS Min % 97.5

Specifications

Chemical Name or Material Acetyl-β-methylcholine chloride
CAS 62-51-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H18ClNO2
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Cl
Formula Weight 195.69
MDL Number MFCD00011817
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents