Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetyl Angiotensinogen (1-14), Porcine

For cellular and molecular biology applications

Specifications

Product Type Acetyl Angiotensinogen (1-14), porcine
Cross Reactivity Porcine
Molecular Weight 1801.08
Storage Requirements -30° to -10°C
Sequence Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66071LB0

 
alfa aesar™
J66071LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66071MCR

 
alfa aesar™
J66071MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Acetyl Angiotensinogen (1-14), porcine
Cross Reactivity Porcine
Molecular Weight 1801.08
Storage Requirements -30° to -10°C
Sequence Ac-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser
Format Solid
Regulatory Status RUO
Purification Method HPLC