missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl Acetone (Certified ACS), Fisher Chemical

Manufacturer:  Fisher Chemical BPA25500

Catalog No. A25-500




Description

Description

Specifications

Specifications

Acetyl Acetone
33°C
Glass Bottle
133°C
-23°C
6
C5H8O2
MFCD00008787
2,4-dioxopentane, 2,4-pentadione, 2,4-pentanedione, acac, acetoacetone, acetyl acetone, acetylacetone, diacetylmethane, pentan-2,4-dione, pentanedione
CC(=O)CC(C)=O
100.12
CHEBI:14750
Liquid
0.975g/cm³
Pass Test
0.972 to 0.978 (at 20°C)
Colorless
500mL
123-54-6
CH3COCH2COCH3
2310
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
31261
100.12
Certified

Chemical Identifiers

123-54-6
100.12
YRKCREAYFQTBPV-UHFFFAOYSA-N
31261
pentane-2,4-dione
C5H8O2
MFCD00008787
2,4-dioxopentane, 2,4-pentadione, 2,4-pentanedione, acac, acetoacetone, acetyl acetone, acetylacetone, diacetylmethane, pentan-2,4-dione, pentanedione
CHEBI:14750
CC(=O)CC(C)=O
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 3: Flammable Liquid

SDS Documents
Product Certifications