Please login to your online account to display your discounted pricing

Acetoxymethylpolystyrene resin, 1%crosslinked with DVB,1.1-1.5mmol/g,100-200 mesh, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  349490250

 View more versions of this product

Catalog No. AC349490250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 25g
Color Off-white to beige
Additional Information crosslinking: 1% DVB
Chemical Name or Material Acetoxymethylpolystyrene resin
Name Note 1% Crosslinked with DVB, 1.1-1.5 mmol/g
MDL Number MFCD02685746
Loading 1.1 to 1.5mmol/g
Mesh Size 100 to 200 mesh

Specifications

Chemical Name or Material Acetoxymethylpolystyrene resin
Name Note 1% Crosslinked with DVB, 1.1-1.5 mmol/g
Physical Form Beads
MDL Number MFCD02685746