Please login to your online account to display your discounted pricing

Acetoxymethylpolystyrene resin, 1%crosslinked with DVB,1.1-1.5mmol/g,100-200 mesh, Acros Organics

Chemical Identifiers

Chemical Name or Material Acetoxymethylpolystyrene resin
Name Note 1% Crosslinked with DVB, 1.1-1.5 mmol/g
Physical Form Beads
MDL Number MFCD02685746
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC349490250

  SDS
thermo fisher scientific
349490250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC349490050

  SDS
thermo fisher scientific
349490050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Beads
Quantity 25g
Color Off-white to beige
Additional Information crosslinking: 1% DVB
Chemical Name or Material Acetoxymethylpolystyrene resin
Name Note 1% Crosslinked with DVB, 1.1-1.5 mmol/g
MDL Number MFCD02685746
Loading 1.1 to 1.5mmol/g
Mesh Size 100 to 200 mesh

for solid support synthesis