Please login to your online account to display your discounted pricing

Acetoxymethylpolystyrene resin, 1% crossl. with DVB,0.5-0.8mmol/g,100-200 mesh, Acros Organics

Chemical Identifiers

Chemical Name or Material Acetoxymethylpolystyrene resin
Name Note 1% Crosslinked with DVB, 0.5-0.8 mmol/g
Physical Form Beads
MDL Number MFCD02685746
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC349480250

  SDS
thermo fisher scientific
349480250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Beads
Quantity 25g
Color Off-white
Chemical Name or Material Acetoxymethylpolystyrene resin
Name Note 1% Crosslinked with DVB, 0.5-0.8 mmol/g
MDL Number MFCD02685746
Loading 0.5 to 0.8mmol/g
Mesh Size 100 to 200 mesh
Additional Information crosslinking: 1% DVB

for solid support synthesis