Learn More
Acetoxyacetyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 370070250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetoxyacetyl chloride | |
13831-31-7 | |
100.0 | |
64°C to 68°C (25.0mmHg) | |
Authentic | |
Glass bottle | |
1.4255 to 1.4275 | |
MFCD00011535 | |
1.27 | |
HZDNNJABYXNPPV-UHFFFAOYSA-N | |
(2-chloro-2-oxoethyl) acetate | |
26297 | |
97% |
97% | |
96.0 | |
1.2700g/mL | |
71°C | |
96% min. (GC) | |
C4H5ClO3 | |
CH3CO2CH2COCl | |
25g | |
acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920 | |
CC(=O)OCC(=O)Cl | |
136.53 | |
136.53 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 237-542-4
RUO â Research Use Only