missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetovanillone, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC102425000

 View more versions of this product

Catalog No. AC102425000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Acetovanillone
Glass bottle
295.0°C to 300.0°C
112.0°C to 116.0°C
498-02-2
100.0
C9H10O3
MFCD00008747
15, 728
DFYRUELUNQRZTB-UHFFFAOYSA-N
1-(4-hydroxy-3-methoxyphenyl)ethanone
2214
166.18
98%
Authentic
0.5% max. (K.F.)
Yellow to Brown
500g
97.5
98%
HOC6H3(OCH3)COCH3
08, 272
acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon
CC(=O)C1=CC(=C(C=C1)O)OC
166.176
CHEBI:2781
Powder

Chemical Identifiers

498-02-2
166.176
DFYRUELUNQRZTB-UHFFFAOYSA-N
2214
1-(4-hydroxy-3-methoxyphenyl)ethanone
C9H10O3
MFCD00008747
acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon
CHEBI:2781
CC(=O)C1=CC(=C(C=C1)O)OC
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents