Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetonyltriphenylphosphonium chloride, 99%

$52.12 - $512.30

Chemical Identifiers

Chemical Name or Material Acetonyltriphenylphosphonium chloride
CAS 1235-21-8
Molecular Formula C21H20ClOP
Formula Weight 354.82
MDL Number MFCD00011813
Synonym 2-Oxopropyltriphenylphosphonium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1469114

  SDS
alfa aesar™
A1469114
25g Each for $52.12

AAA1469122

  SDS
alfa aesar™
A1469122
100g Each for $123.55

AAA1469136

  SDS
alfa aesar™
A1469136
500g Each for $512.30
Description & Specifications

Specifications

Formula Weight 354.82
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number TA1841000
MDL Number MFCD00011813
CAS 1235-21-8
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Molecular Formula C21H20ClOP
Melting Point 244° to 248°C
Synonym 2-Oxopropyltriphenylphosphonium chloride
Assay 99%
Chemical Name or Material Acetonyltriphenylphosphonium chloride

Applications
Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

Solubility
Soluble in hot water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.