missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile 99.5+%, TCI America™

$22.92 - $33.61

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A006025ML View Documents TCI America
A006025ML
25mL Each for $22.92
 
A0060500ML View Documents TCI America
A0060500ML
500mL Each for $33.61
 
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

82°C
-46°C
75-05-8
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
Clear Liquid at 20°C
Acetonitrile
Colorless
25mL
C2H3N
1648
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
≥99.5% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1508

RTECSNumber : AL7700000

TSCA : Yes