missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, Spectrophotometric Grade, 99.7%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32470M1

 View more versions of this product

Catalog No. AA32470M1



Specifications

Specifications

Acetonitrile
75-05-8
>99.7%
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
Sprectrophotometric
97 hPa (120mm Hg) at 20°C
4 x 1L
Meets ACS spectrophotometric requirements. Filtered through 0.2μfilters
Colorless liquid
C2H3N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
Liquid
Pungent
Colorless

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
Safety and Handling

Safety and Handling

H225-H302-H312-H332-H319: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-835-2

SDS Documents