missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile (Pesticide), Fisher Chemical

Manufacturer:  Fisher Chemical FLA9994

Catalog No. A999-4




Specifications

Specifications

Acetonitrile
Vapor Pressure: 97mbar at 20°C
C2H3N
MFCD00001878
acetonitril
CC#N
41.053
CHEBI:38472
Liquid
0.781g/cm³
Glass Bottle
97mbar at 20°C
0.36 cP at 20°C
81°C
-46°C
75-05-8
Also meets ACS specifications
CH3CN
1648; Colorless
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
Pesticide
Pass Test
10 ng/L max.
12.8°C (55°F)
>500°C
Colorless
4L

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
acetonitril
CHEBI:38472
CC#N
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

SDS Documents
Accessories