Learn More
Acetonitrile, HPLC Grade (Far UV), 99.8+%, Thermo Scientific Chemicals
Suitable For Far UV Analysis
Supplier: Thermo Scientific Chemicals 42880K2
Description
It is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetonitrile | |
75-05-8 | |
0.786 | |
1 L | |
525°C (977°F) | |
Liquid | |
>99.8% | |
MFCD00001878 | |
1648; Colorless | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 |
°C to 48°C to°C to 45°C | |
Colorless | |
80°C to 82°C | |
5°C (41°F) | |
Pungent | |
1.344 | |
C2H3N | |
73mm Hg at 20°C | |
741857 | |
Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons. | |
CC#N | |
41.053 | |
CHEBI:38472 | |
HPLC |
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Safety and Handling
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 200-835-2
RTECSNumber : AL7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only