Learn More
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 22927M1
Description
Acetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetonitrile | |
75-05-8 | |
Colorless liquid | |
80°C to 82°C | |
5°C (41°F) | |
Liquid | |
>99.7% min. | |
MFCD00001878 | |
1648;Colorless | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 |
-48°C to -45°C | |
Colorless | |
0.786 | |
4 x 1 L | |
Pungent | |
1.344 | |
C2H3N | |
97 hPa (73mm Hg) at 20°C | |
741857 | |
Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons. | |
CC#N | |
41.053 | |
CHEBI:38472 | |
HPLC |
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Safety and Handling
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 200-835-2
RTECSNumber : AL7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only