Learn More
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 22927M6
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetonitrile | |
75-05-8 | |
80°C to 82°C | |
5°C (41°F) | |
Liquid | |
C2H3N | |
UN1648 | |
14,70 | |
Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons. | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.7% |
-48°C to -45°C | |
0.786 | |
4 x 4 L | |
Pungent | |
1.344 | |
MFCD00001878 | |
741857 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
HPLC |
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Safety and Handling
GHS H Statement
H225-H302-H312-H332-H319;
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
EINECSNumber : 200-835-2
RTECSNumber : AL7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only