missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, HPLC Grade, 99.7+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 22927K7

 View more versions of this product

Catalog No. AA22927K7



Specifications

Specifications

Acetonitrile
C2H3N
UN1648
14,70
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
≥99.7%
0.786
1.3440
80°C to 82°C
4L
75-05-8
MFCD00001878
741857
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
Liquid
HPLC
Pungent
5°C (41°F)
-48°C to -45°C

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H312-H332-H319;
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE

RTECSNumber : AL7700000

TSCA : TSCA

SDS Documents