Please login to your online account to display your discounted pricing

Acetonitrile (HPLC), Fisher Bioreagents™

Submicron Filtered

$96.70 - $193.16

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Purity Grade HPLC Grade
Purity Grade Notes Also meets ACS specifications
Assay ≥99.9%
Assay Percent Range ≥99.9 %
Molecular Formula C2H3N
Linear Formula CH3CN
Formula Weight 41.05
Synonym Ethanenitrile
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP2405SK-1

  SDS
fisher scientific company
BP2405SK1
Glass Bottle 1L Each for $96.70
Case of 6 Each for $580.20

BP2405SK-4

  SDS
fisher scientific company
BP2405SK4
Safe-cote 4L Each for $193.16
Case of 4 Each for $772.64
Description & Specifications

Specifications

Assay ≥99.9%
Melting Point −46°C
Boiling Point 81°C
Formula Weight 41.05
Purity Grade HPLC Grade
Quantity 1L
Merck Index 15, 69
Physical Form Liquid
Color Colorless
Decomposition Information >500°C
Vapor Pressure 97mbar at 20°C
Density 0.781g/cm³
Viscosity 0.36 cP at 20°C
Absorbance 0.01 AU max. at 254nm
Refractive Index 1.3405 to 1.3425 (at 25°C)
Identification Pass Test
CAS 75-05-8
ChemAlert Storage Symbol Red
Purity Grade Notes Also meets ACS specifications
Linear Formula CH3CN
Additional Information Vapor Pressure: 97mbar at 20°C
Titratable Base 0.0006mEq/g max.
Synonym Ethanenitrile
LCMS Gradient Suitability Pass Test
Fluorescence Background Pass Test
Residue after Evaporation 1.0ppm max.
Water 0.01% max.
Packaging Glass Bottle
Assay Percent Range ≥99.9 %
Titratable Acid 0.008mEq/g max.
Health Hazard 2 DANGER!
Chemical Name or Material Acetonitrile
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Molecular Formula C2H3N
Cleanup Symbol Solvent

Also available in recyclable FisherPak™ and NOWPak™ containers.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

DANGER!