Please login to your online account to display your discounted pricing

Acetonitrile, For Spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$279.66 - $582.34

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Molecular Formula CH3CN
Formula Weight 41.05
UN Number 1648
Synonym Methyl cyanide, Ethyl nitrile, Cyanomethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1000161000

 
emd millipore corporation
1000161000
Glass Bottle 1L Each for $279.66

M1000162500

 
emd millipore corporation
1000162500
Glass Bottle 2.5L Each for $582.34
Description & Specifications

Specifications

Boiling Point 81.6°C
Melting Point −45.7°C
Formula Weight 41.05
Quantity 1L
Chemical Name or Material Acetonitrile
Ignition Point 524°C (975°F)
Odor Ether-like
Packaging Glass Bottle
Storage Information 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.
Vapor Pressure 97 hPa (20°C)
Flash Point 2°C (36°F)
Physical Form Liquid
Color Colorless
Molecular Formula CH3CN
CAS 75-05-8
UN Number 1648
Synonym Methyl cyanide, Ethyl nitrile, Cyanomethane
Solubility Information Soluble in water at 20°C
Health Hazard 1 H225: Highly flammable liquid and vapour.
Health Hazard 2 H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Density 0.79g/cm3 at 20°C
Health Hazard 3 H319: Causes serious eye irritation.
Safety and Handling

H225: Highly flammable liquid and vapour.

H319: Causes serious eye irritation.

H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled