Please login to your online account to display your discounted pricing

Acetonitrile, For gas chromatography MS SupraSolv™, EMD Millipore

For gas chromatography MS SupraSolv™

$204.80 - $428.63

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Molecular Formula CH3CN
Formula Weight 41.05
UN Number 1648
Synonym Ethyl nitrile, Cyanomethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1006651000

 
emd millipore corporation
1006651000
Glass Bottle 1L Each for $204.80

M1006652500

 
emd millipore corporation
1006652500
Glass Bottle 2.5L Each for $428.63
Description & Specifications

Specifications

Boiling Point 81.6°C
Melting Point −45.7°C
Formula Weight 41.05
Quantity 1L
Description SupraSolv™ MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Health Hazard 2 H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
Chemical Name or Material Acetonitrile
Ignition Point 524°C (975°F)
Odor Ether-like
Packaging Glass Bottle
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
Vapor Pressure 97 hPa (20°C)
Flash Point 2°C (36°F)
Physical Form Liquid
Color Colorless
Molecular Formula CH3CN
CAS 75-05-8
UN Number 1648
Synonym Ethyl nitrile, Cyanomethane
Solubility Information Soluble in water at 20°C
Health Hazard 1 H225: Highly flammable liquid and vapour.
Density 0.79g/cm3 at 20°C
Health Hazard 3 H319: Causes serious eye irritation.
Safety and Handling

H225: Highly flammable liquid and vapour.

H319: Causes serious eye irritation.

H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled