Please login to your online account to display your discounted pricing

Acetonitrile, For gas chromatography ECD and FID SupraSolv™, EMD Millipore

For gas chromatography ECD and FID SupraSolv™

$200.49 - $419.57

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Molecular Formula CH3CN
Formula Weight 41.05
UN Number 1648
Synonym Methyl cyanide, Ethyl nitrile, Cyanomethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1000171000

 
emd millipore corporation
1000171000
Glass Bottle 1L Each for $200.49

M1000172500

 
emd millipore corporation
1000172500
Glass Bottle 2.5L Each for $419.57
Description & Specifications

Specifications

Chemical Name or Material Acetonitrile
CAS 75-05-8
Health Hazard 1 H225: Highly flammable liquid and vapour.
Health Hazard 2 H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
Health Hazard 3 H319: Causes serious eye irritation.
Physical Form Liquid
Molecular Formula CH3CN
Formula Weight 41.05
UN Number 1648
Synonym Methyl cyanide, Ethyl nitrile, Cyanomethane
Solubility Information Soluble in water at 20°C
Description SupraSolv™ ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Density 0.79g/cm3 at 20°C
Ignition Point 524°C (975°F)
Odor Ether-like
Packaging Glass Bottle
Refractive Index 1.34
Storage Information 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.
Vapor Pressure 97 hPa (20°C)
Flash Point 2°C (36°F)
Boiling Point 81.6°C
Color Colorless
Melting Point −45.7°C
Quantity 1L
Safety and Handling

H225: Highly flammable liquid and vapour.

H319: Causes serious eye irritation.

H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled