missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile-d3, for NMR, 99% atom D, ACROS Organics™

$104.88 - $415.26

Chemical Identifiers

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.071
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
Synonym 2h3 acetonitrile
PubChem CID 123151
IUPAC Name 2,2,2-trideuterioacetonitrile
SMILES CC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC166230100 SDS View Documents Thermo Fisher Scientific
166230100
10mL Glass bottle Each for $104.88
 
AC166230250 SDS View Documents Thermo Fisher Scientific
166230250
25mL Glass bottle Each for $229.22
 
AC166230500 SDS View Documents Thermo Fisher Scientific
166230500
50mL Glass bottle Each for $415.26
 
Description

Description

For NMR
Specifications

Chemical Identifiers

2206-26-0
44.071
WEVYAHXRMPXWCK-FIBGUPNXSA-N
123151
CC#N
C2H3N
MFCD00001881
2h3 acetonitrile
2,2,2-trideuterioacetonitrile

Specifications

98.5% min.
0.1% max. (Coulometric)
10mL
0.39 mPa.s (20°C)
79.0°C to 81.0°C
C2H3N
MFCD00001881
2°C
2h3 acetonitrile
CC#N
44.071
44.07
NMR
Glass bottle
Colorless
2206-26-0
0.8440g/mL
-46.0°C
CD3CN
02, IV, 428
1.3430 to 1.3450
WEVYAHXRMPXWCK-FIBGUPNXSA-N
2,2,2-trideuterioacetonitrile
123151
Liquid
Acetonitrile-d3, 99 atom%D
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger