missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile-D3 deuteration degree min. 99.96%, for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

Manufacturer:  Emd Millipore Corporation 1137530001

 View more versions of this product

Catalog No. M1137530001


 

Specifications Description & Specifications

Specifications

Acetonitrile-D3
Ether-like
525°C (977°F)
0.39mPa&middnot;s (20°C)
1mL
Colorless
44.07
2206-26-0
Glass ampoule
97 hPa at 20°C (68°F)
Liquid
99% min.
CD3CN

Chemical Identifiers

2206-26-0
99% min.
44.07
Liquid
CD3CN
Acetonitrile-D3