Please login to your online account to display your discounted pricing

Acetonitrile-D3 deuteration degree min. 99.96%, for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$47.08 - $245.08

Chemical Identifiers

Chemical Name or Material Acetonitrile-D3
CAS 2206-26-0
Physical Form Liquid
Assay Percent Range 99% min.
Molecular Formula CD3CN
Formula Weight 44.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1137530009

 
emd millipore corporation
1137530009
Glass ampoule 10 x 0.75mL Each for $245.08

M1137530001

 
emd millipore corporation
1137530001
Glass ampoule 1mL Each for $47.08
Description & Specifications

Specifications

Chemical Name or Material Acetonitrile-D3
CAS 2206-26-0
Odor Ether-like
Packaging Glass ampoule
Ignition Point 525°C (977°F)
Vapor Pressure 97 hPa at 20°C (68°F)
Viscosity 0.39mPa&middnot;s (20°C)
Physical Form Liquid
Quantity 10 x 0.75mL
Assay Percent Range 99% min.
Color Colorless
Molecular Formula CD3CN
Formula Weight 44.07