Please login to your online account to display your discounted pricing

Acetonitrile-D3 deuteration degree min. 99.96%, for NMR spectroscopy, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 10 x 0.75mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  1137530009

 View more versions of this product

Catalog No. M1137530009

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Acetonitrile-D3
CAS 2206-26-0
Odor Ether-like
Packaging Glass ampoule
Ignition Point 525°C (977°F)
Vapor Pressure 97 hPa at 20°C (68°F)
Viscosity 0.39mPa&middnot;s (20°C)
Physical Form Liquid
Quantity 10 x 0.75mL
Assay Percent Range 99% min.
Color Colorless
Molecular Formula CD3CN
Formula Weight 44.07

Specifications

Chemical Name or Material Acetonitrile-D3
CAS 2206-26-0
Physical Form Liquid
Assay Percent Range 99% min.
Molecular Formula CD3CN
Formula Weight 44.07