Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetonitrile-d3, 100% (Isotopic)

Chemical Identifiers

Chemical Name or Material Acetonitrile-d3
CAS 2206-26-0
Name Note Isotopic
Formula Weight 44.08
MDL Number MFCD00001881
UN Number UN1648
Merck Index 14,70
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42266KV

  SDS
alfa aesar™
42266KV
10 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid In Prescored Ampoules, (0.75Ml/ampoule)
Formula Weight 44.08
Chemical Name or Material Acetonitrile-d3
Sensitivity Hygroscopic
Density 0.844g/mL
Name Note Isotopic
Boiling Point 81°C
Assay 100%
Quantity 10 each
Color Colorless
Flash Point 5° (42°F)
MDL Number MFCD00001881
CAS 2206-26-0
UN Number UN1648
Merck Index 14,70
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H302-H312-H332-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Odor Pungent
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H302-H312-H332-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.