missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ Acetonitrile, CHROMASOLV™, ≥99.9% (GC), for LC-NMR, Honeywell Riedel-de Haën™

CHROMASOLV™ NMR, for LC-NMR,≥99.9% (GC)

Manufacturer:  Honeywell Riedel-de Haen™ 34955500ML

 View more versions of this product

Catalog No. 6000254



Description

Description

  • Solvents for LC-NMR applications
Specifications

Specifications

Acetonitrile
C2H3N
MFCD00001878
741857
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05g/mol
CHROMASOLV™
0.16
72.8 mmHg (20°C)
2°C (35.6°F)
500mL
75-05-8
CH3CN
UN1648
acetonitril
CC#N
41.053
CHEBI:38472
≥99.9% (GC)
0.786g/mL at 25°C
973°F
1.41 (vs air)
Colorless

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
acetonitril
CHEBI:38472
CC#N
Safety and Handling

Safety and Handling

P210-P280-P305 + P351 + P338

RTECSNumber : AL7700000

Documents