Please login to your online account to display your discounted pricing

Acetonitrile ChromAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Molecular Formula CH3CN
Formula Weight 41.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK28561

 
avantor performance
285606
Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK28564

 
avantor performance
285610
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MKH0764

 
avantor performance
H07610
SAFEMOR 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 41.05
Quantity 1L
Chemical Name or Material Acetonitrile
Packaging Glass
Molecular Formula CH3CN
CAS 75-05-8