Please login to your online account to display your discounted pricing

Acetonitrile, BAKER ANALYZED™ ULTRA LC/MS, J.T.Baker™

For cutting-edge LC/MS applications

$357.08 - $570.72

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Purity Grade BAKER ANALYZED™ ULTRA LC/MS
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula CH3CN
Formula Weight 41.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002173

 
avantor performance
985302
Narrow Mouth Flint 1L Each for $357.08

02002172

 
avantor performance
985301
Narrow Mouth Flint 1L Each for $570.72
Description & Specifications

Specifications

Formula Weight 41.05
Purity Grade Notes Meets ACS reagent chemical requirements
Quantity 1L
Water ≤100ppb
Sodium (Na) ≤50.0ppb
Iron (Fe) ≤5.0ppb
Assay ≥99.9%
Packaging Narrow Mouth Flint
CAS 75-05-8
Molecular Formula CH3CN
Chemical Name or Material Acetonitrile
Flash Point 2.00°C
Tin (Sn) ≤5.0ppb
Specific Gravity 1L = 0.79kg
Nickel (Ni) ≤5.0ppb
Lead (Pb) ≤5.0ppb
Calcium (Ca) ≤25.0ppb
Manganese (Mn) ≤5.0ppb
Residue after Evaporation ≤0.8ppm
Purity Grade BAKER ANALYZED™ ULTRA LC/MS
Magnesium (Mg) ≤10.0ppb
Aluminum (Al) ≤25.0ppb
Chromium (Cr) ≤5.0ppb
Cobalt (Co) ≤5.0ppb
Copper (Cu) ≤5.0ppb

    For use in LC/MS and UHPLC