Please login to your online account to display your discounted pricing

Acetonitrile, BAKER ANALYZED™ HPLC ULTRA Gradient Solvent, J.T.Baker™

$321.02 - $901.00

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Molecular Formula CH3CN
Formula Weight 41.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002175

 
avantor performance
901702
Narrow Mouth Amber Glass Bottle 1L Each for $321.02

02002182

 
avantor performance
901733
Narrow Mouth PVC Coated Amber Glass Bottle 4L Each for $901.00
Description & Specifications

Specifications

Formula Weight 41.05
Purity Grade HPLC
Chemical Name or Material Acetonitrile
Flash Point 2°C (35.8°F)
Molecular Formula CH3CN
CAS 75-05-8
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L