Please login to your online account to display your discounted pricing

Acetonitrile, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$259.02 - $2,468.97

Chemical Identifiers

Chemical Name or Material Acetonitrile
CAS 75-05-8
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CH3CN
Formula Weight 41.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002179

 
avantor performance
901103
Narrow Mouth Amber Glass Bottle 4L Each for $2,468.97

02002184

 
avantor performance
901101
Narrow Mouth Amber Glass Bottle 500mL Each for $259.02
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 41.05
Chemical Name or Material Acetonitrile
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4L
Specific Gravity 1L = 0.79kg
Flash Point 2.00°C
Molecular Formula CH3CN
CAS 75-05-8