Please login to your online account to display your discounted pricing

Acetonitrile, anhydrous, >99.8%, Alfa Aesar

$39.78 - $104.86

Chemical Identifiers

Chemical Name or Material Acetonitrile, anhydrous
Name Note Water <100ppm
CAS 75-05-8
Physical Form Liquid
Assay Percent Range >99.8%
Molecular Formula CH3CN
Formula Weight 41.05
MDL Number MFCD00001878
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43166K2

  SDS
alfa aesar
43166K2
1L Each for $104.86

AA43166AK

  SDS
alfa aesar
43166AK*
250mL Each for $39.78
Description & Specifications

Specifications

Formula Weight 41.05
Quantity 1L
EINECS Number 200-835-2
Physical Form Liquid
Density 0.786
Chemical Name or Material Acetonitrile, anhydrous
Name Note Water <100ppm
Assay Percent Range >99.8%
Molecular Formula CH3CN
MDL Number MFCD00001878
CAS 75-05-8