Please login to your online account to display your discounted pricing

Acetonitrile, ACS, EMD Millipore

$5,850.40 - $5,850.40

Chemical Identifiers

Chemical Name or Material Acetonitrile
Name Note For ACS analysis
CAS 75-05-8
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula CH3CN
Formula Weight 41.05
Synonym Methyl Cyanide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MAX014920

 
emd millipore corporation
AX014920
Metal Drum 200L Each for $5,850.40
Description & Specifications

Specifications

Formula Weight 41.05
Physical Form Liquid
Color Clear colorless
Quantity 200L
Packaging Metal Drum
Chemical Name or Material Acetonitrile
Assay Percent Range 99.5% min.
Name Note For ACS analysis
CAS 75-05-8
Molecular Formula CH3CN
Synonym Methyl Cyanide