Please login to your online account to display your discounted pricing

Acetonitrile ACS AR, Macron Fine Chemicals™


Quantity 500mL
Packaging Glass

Manufacturer: avantor performance  004304

 View more versions of this product

Catalog No. MK0043500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 41.05
Quantity 500mL
Chemical Name or Material Acetonitrile
Packaging Glass
Molecular Formula CH3CN
CAS 75-05-8

Specifications

Chemical Name or Material Acetonitrile
CAS 75-05-8
Purity Grade ACS Reagent Grade
Molecular Formula CH3CN
Formula Weight 41.05