Please login to your online account to display your discounted pricing

Acetonitrile, 99.9%, for residue analysis, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  326750010

 View more versions of this product

Catalog No. AC326750010

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Acetonitrile
Name Note Tested for Trace Analysis of polyaromatic Hydrocarbons
CAS 75-05-8
CAS Min % 99.85
Health Hazard 1 Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Acetic Acid 0.001% max.
RTECS Number AL7700000
EINECS Number 200-835-2
TSCA TSCA
Purity Grade For residue analysis
Physical Form Liquid
Assay ≥99.9%
Assay Percent Range 99.9%
Molecular Formula C2H3N
Linear Formula CH3CN
Formula Weight 41.04
MDL Number MFCD00001878
Beilstein 02, 183
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Synonym AN
Density 0.7810g/mL
Packaging Glass bottle
Residue after Evaporation 0.0005% max.
Water 0.05% max. (Coulometric)
Color Clear
Quantity 1L

Specifications

Chemical Name or Material Acetonitrile
Name Note Tested for Trace Analysis of polyaromatic Hydrocarbons
CAS 75-05-8
CAS Min % 99.85
CAS Max % 100.0
Purity Grade For residue analysis
Assay ≥99.9%
Assay Percent Range 99.9%
Molecular Formula C2H3N
Linear Formula CH3CN
Formula Weight 41.04
MDL Number MFCD00001878
Synonym AN
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

SDS & Safety Documents