Learn More
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methylkyanid, methane, cyano, methyl cyanide, mecn, methanecarbonitrile, ethanenitrile, ethyl nitrile, cyanure de methyl, cyanomethane, acetonitrile, 100mL, 0.002 % max., Vapor Pressure: 97mbar at 20 deg.C, 99.9+%, 02, 183, 81.0 deg.C to 82.0 deg.C
Supplier: Thermo Scientific Chemicals 326811000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetonitrile | |
75-05-8 | |
0.7810g/mL | |
100 mL | |
Authentic | |
1.3430 to 1.3450 | |
99.9+% | |
C2H3N | |
MFCD00001878 | |
0.36 mPa.s (20°C) | |
02, 183 | |
15, 69 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.9% |
-46.0°C | |
>500°C | |
81.0°C to 82.0°C | |
2°C | |
Liquid | |
0.0005 % max. | |
0.781 | |
CH3CN | |
Vapor Pressure: 97mbar at 20°C | |
0.001% Max. (Coulometric) | |
02,13; 12,450; 13,156; 15,1 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
Extra Dry |
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only