missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™

$141.98 - $141.98

Chemical Identifiers

CAS 75-05-8
Assay Percent Range 99.9%
Molecular Formula C2H3N
Linear Formula CH3CN
Formula Weight 41.04
MDL Number MFCD00001878
Synonym AN
Chemical Name or Material Acetonitrile
Assay ≥99.9%
Grade Extra Dry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC610220010 SDS Fisher Scientific Acros
610220010
AcroSeal™ Glass Bottle in Tin Can 1L Each for $141.98
Specifications Description & Specifications

Chemical Identifiers

75-05-8
C2H3N
41.04
AN
≥99.9%
99.9%
CH3CN
MFCD00001878
Acetonitrile
Extra Dry

Specifications

75-05-8
99.9%
CH3CN
MFCD00001878
02, 183
15, 69
0.7810g/mL
Conforms to reference
1ppm max.
2°C (35.8°F)
81° to 82°C
1L
≥99.9%
Vapor Pressure: 97mbar at 20°C
C2H3N
41.04
1648; Colorless
02,13; 12,450; 13,156; 15,1
AN
Pass Test
AcroSeal™ Glass Bottle in Tin Can
50ppm max.
>500°C
−46°C
Acetonitrile
Extra Dry

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-835-2

RTECSNumber : AL7700000

TSCA : TSCA