missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, 99.8%, for HPLC, ACROS Organics™

$199.26 - $325.79

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym acetonitril
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC268270010 SDS View Documents Fisher Scientific Acros
268270010
1L Glass bottle Each for $199.26
 
AC268270025 SDS View Documents Thermo Fisher Scientific
268270025
2.5L Glass bottle Each for $325.79
 
Description

Description

For HPLC
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
acetonitril
CHEBI:38472
CC#N

Specifications

(1cm cell vs HPLC−grade water)
(1cm cell vs HPLC-grade water)
Vapor Pressure: 97mbar at 20°C
C2H3N
MFCD00001878
02, 183
15, 69
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
CC#N
41.053
CHEBI:38472
Liquid
HPLC
Yes
1.3430 to 1.3450
0.781
2°C (35.8°F)
>500°C
Undesignated
1L
75-05-8
0.0008 meq/g max.
99.8%
CH3CN
1648; Colorless
02,13; 12,450; 13,156; 15,1
acetonitril
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
99.8%
0.7810g/mL
Glass bottle
0.0002% max.
0.05% max.
0.36 mPa.s (20°C)
81.0°C to 82.0°C
-46.0°C
Acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

Danger

EINECSNumber : 200-835-2

RTECSNumber : AL7700000

TSCA : TSCA