Learn More
Acetonitrile, 99.8%, for HPLC, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 268270040
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCSpecifications
Acetonitrile | |
75-05-8 | |
0.0008 meq/g max. | |
0.7810g/mL | |
4 L | |
Glass Bottle | |
1.3430 to 1.3450 | |
99.8% | |
C2H3N | |
MFCD00001878 | |
0.36 mPa.s (20°C) | |
0.05% max. | |
02,13; 12,450; 13,156; 15,1 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
HPLC |
-46.0°C | |
(1cm cell vs HPLC−grade water), 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.01 max. at 250nm, 0.015 max. at 240nm, 0.05 max. at 230nm | |
>500°C | |
81.0°C to 82.0°C | |
2°C (35.8°F) | |
Liquid | |
0.0002% max. | |
0.781 | |
CH3CN | |
Vapor Pressure: 97mbar at 20°C | |
1648; Colorless | |
02, 183 | |
15, 69 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether | |
CC#N | |
41.053 | |
CHEBI:38472 | |
99.8% |
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 200-835-2
RTECSNumber : AL7700000
TSCA : TSCA
RUO – Research Use Only