missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, 99.5%, for analysis, ACROS Organics™

$1,828.46 - $1,828.46

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym acetonitril, cyanomethane, cyanure de methyl, ethanenitrile, ethyl nitrile, mecn, methane, cyano, methanecarbonitrile, methyl cyanide, methylkyanid
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC258560250
SDS View Documents
Thermo Fisher Scientific
AC258560250
25L Metal drum
Each for $1,828.46
 
Description

Description

For analysis
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
acetonitril, cyanomethane, cyanure de methyl, ethanenitrile, ethyl nitrile, mecn, methane, cyano, methanecarbonitrile, methyl cyanide, methylkyanid
CHEBI:38472
CC#N

Specifications

75-05-8
100.0
C2H3N
MFCD00001878
02,13; 12,450; 13,156; 15,1
acetonitril, cyanomethane, cyanure de methyl, ethanenitrile, ethyl nitrile, mecn, methane, cyano, methanecarbonitrile, methyl cyanide, methylkyanid
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
CHEBI:38472
Liquid
Analysis
0.7810g/mL
0.001% max.
2ppm max.
Metal drum
1.3430 to 1.3450
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
0.781
0.1% max. (Coulometric)
0.36 mPa.s (20°C)
Undesignated
25L
99.4
99.5%
CH3CN
02, 183
15, 69
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
CC#N
41.053
1ppm max.
41.05
99.5%
2ppm max.
0.005% max.
2ppm max.
0.5ppm max.
1ppm max.
0.005% max.
5ppm max.
2ppm max.
2°C
81.0°C to 82.0°C
-46.0°C
Acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS