Please login to your online account to display your discounted pricing

Acetonitrile, 0.1% Trifluoroacetic Acid; Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Function tested and optimized for LC/MS applications

$375.70 - $1,135.57

Chemical Identifiers

Chemical Name or Material Acetonitrile
Name Note 0.1% Trifluoroacetic Acid; Filtered through a 0.2 micron filter
Purity Grade BAKER ANALYZED™ LC-MS Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002170

 
avantor performance
983503
Narrow Mouth Amber Glass Bottle 4L Each for $1,135.57

02002169

 
avantor performance
983502
Narrow Mouth Amber Glass Bottle 1L Each for $375.70
Description & Specifications

Specifications

Molecular Formula C2H3N
Linear Formula CH3CN
UN Number 1648; Colorless
Chemical Name or Material Acetonitrile
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.40kg
Name Note 0.1% Trifluoroacetic Acid; Filtered through a 0.2 micron filter
Purity Grade BAKER ANALYZED™ LC-MS Reagent
Flash Point 2.00°C
Quantity 4L

    For use in liquid chromatography and mass spectrometry; For laboratory, research, or manufacturing use