Please login to your online account to display your discounted pricing

Acetone (UltimAR), Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade UltimAR
Molecular Formula CH3COCH3
Formula Weight 58.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MKH4511

 
avantor performance
H45106
Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MKH4514

 
avantor performance
H45110
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade UltimAR
Molecular Formula CH3COCH3
Formula Weight 58.08
Packaging Glass
Quantity 1L